4-Ethoxy-2,3-difluorobenzyl alcohol ≥97%
|
Warning
|
ALFAH32868.03EA
113
SGD
ALFAH32868.03
ALFAH32868.06
4-Ethoxy-2,3-difluorobenzyl alcohol ≥97%
4-Ethoxy-2,3-difluorobenzyl alcohol
Formula:
C₉H₁₀F₂O₂ MW: 188,17 g/mol Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258704 CAS Number: 293299-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...