Would you like to log in again?
Thermometer adapters
Supplier: Corning Life Sciences
Thermometer adapters are designed with a ground joint at one end. The other end is tooled to accommodate a thermometer. Adapters are supplied with a rubber thermometer holder.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Cone size Pk VWR Catalog Number Quantity
14/20 NS 1 CORN8821-14
Supplier Article Number:: 8821-14
19/22 NS 1 CORN8821-19
Supplier Article Number:: 8821-19
24/40 NS 1 CORN8821-24
Supplier Article Number:: 8821-24
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty