Would you like to log in again?
Enzyme-active powdered detergent, Tergazyme™
Supplier: ALCONOX
Concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning. Removes proteinaceous soils, tissue, blood, and body fluids from glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fibreglass. Ideal as a cleaning agent in reverse osmosis and ultra-filtration equipment used in dairy, biotechnology, and pollution processing. Also for presoaking hard to remove stains.
  • Free-rinsing formula leaves no interfering residue
  • Designed to replace corrosive acids and hazardous solvents
  • Biodegradable and non corrosive
  • Dilute 1:100
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Enzyme-active powdered detergent, Tergazyme™ 1,8 kg ALCO1304
Supplier Article Number:: 1304
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty