Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer, potassium phosphate, 0,5M solution, pH 7,2

Supplier: Alfa Aesar
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401



 

ORDER

Expand All / Collapse All
Description Pack type Pk VWR Catalog Number Quantity
Potassium phosphate, 0,5M buffer solution, pH 7,2 Glass bottle 500 ml ALFAJ60326.AP
Supplier Article Number:: J60326.AP
Potassium phosphate, 0,5M buffer solution, pH 7,2 Glass bottle 250 ml ALFAJ60326.AK
Supplier Article Number:: J60326.AK

SPECIFICATIONS

CAS No. 7778-77-0
Physical form Liquid
MDL No. MFCD00011401
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty