Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride ≥95%

Supplier: Alfa Aesar
Warning
Synonyms: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride



Formula: C₁₂H₁₈BClN₂O₃
MW: 284,55
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD09027217
CAS Number: 913835-43-5

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH53296.03
Supplier Article Number:: H53296.03
Glass bottle for solids 250 mg ALFAH53296.MD
Supplier Article Number:: H53296.MD
Expand All / Collapse All

SPECIFICATIONS

Melting point 187-189°
Formula weight 284.55
CAS No. 913835-43-5
Purity 95%
Molecular formula C12H17BN2O3.HCl
MDL No. MFCD09027217
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty