Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Furfurylcarbamoyl)phenylboronic acid ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 850567-27-0
Molecular Formula: C12H12BNO4
Formula Weight: 245.04
Melting Point: 216-220°
MDL No.: MFCD04115700

 
  Formula: C₁₂H₁₂BNO₄
MDL Number: MFCD04115700
CAS Number: 850567-27-0
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH52992.03
Glass bottle for solids 250 mg ALFAH52992.MD
Expand All / Collapse All

SPECIFICATIONS

Melting Point 216-220°
Formula Weight 245.04
CAS No. 850567-27-0
Purity 97%
Molecular Formula C12H12BNO4
MDL No. MFCD04115700
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty