Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid ≥97%

Supplier: Alfa Aesar
Warning
Synonyms: 4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid



Formula: C₁₇H₁₇BN₂O₃
MW: 308,14
Storage Temperature: Ambient
MDL Number: MFCD08689479
CAS Number: 380499-66-1

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg ALFAH52632.MD
Supplier Article Number:: H52632.MD
Expand All / Collapse All

SPECIFICATIONS

Formula weight 308.14
CAS No. 380499-66-1
Purity 97%
Molecular formula C17H17BN2O3
MDL No. MFCD08689479
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty