3,6-Dimethoxypyridazine-4-carboxylic acid 96%
Supplier: Thermo Scientific
|
Warning
|
ALFAH26877.03EA
0
SGD
ALFAH26877.03
3,6-Dimethoxypyridazine-4-carboxylic acid 96%
3,6-Dimethoxypyridazine-4-carboxylic acid
Formula:
C₇H₈N₂O₄ MW: 184,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426073 CAS Number: 89694-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...