4-Bromo-3-methylbenzenesulphonamide ≥97%

Warning

ALFAH32894.06EA 558 SGD
ALFAH32894.06
4-Bromo-3-methylbenzenesulphonamide ≥97%
4-Bromo-3-methylbenzenesulphonamide
Formula: C₇H₈BrNO₂S
MW: 250,11 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD0394591
CAS Number: 879487-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR