Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde ≥97%

Supplier: Alfa Aesar
Warning



Formula: C₉H₈ClFO₂
MW: 202,61
Storage Temperature: Ambient
MDL Number: MFCD22682805
CAS Number: 1823049-90-6

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH33065.03
Supplier Article Number:: H33065.03
Glass bottle for solids 250 mg ALFAH33065.MD
Supplier Article Number:: H33065.MD
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty