3-Chloro-2-fluoro-6-(trifluoromethyl)aniline ≥97%
|
Danger
|
ALFAH32861.MDEA
72.7
SGD
ALFAH32861.MD
ALFAH32861.03
3-Chloro-2-fluoro-6-(trifluoromethyl)aniline ≥97%
3-Chloro-2-fluoro-6-(trifluoromethyl)aniline
Formula:
C₇H₄ClF₄N MW: 213,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519381 CAS Number: 1092461-38-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...