3,5-Difluoro-4-ethoxybenzonitrile ≥97%

Danger

Synonyms: 4-Ethoxy-3,5-difluorobenzonitrile

ALFAH31715.03EA 150 SGD
ALFAH31715.03 ALFAH31715.06
3,5-Difluoro-4-ethoxybenzonitrile ≥97%
3,5-Difluoro-4-ethoxybenzonitrile
Formula: C₉H₇F₂NO
MW: 183,15 g/mol
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD09258687
CAS Number: 1017779-27-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR