2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile ≥97%
|
Danger
|
ALFAH33354.03EA
205
SGD
ALFAH33354.03
ALFAH33354.06
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile ≥97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile
Formula:
C₉H₅F₄NO MW: 219,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201044 CAS Number: 1373920-99-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...