General incubators, natural convection
VWRU414004-612
General incubators, natural convection
Natural convection general incubators combine microprocessor controlled temperature with state-of-the-art design that delivers a true naturally-occurring gravity convection. Designed for convenient and safe use in many applications, such as animal & plant cell culture, constant temperature, and germination testing.
  • Advanced, adaptive, microprocessor control system with rotatory knob resulting in superior temperature accuracy
  • High-quality LCD display with back-light
  • RS232C interface for remote monitoring and controlling with PC
  • Over temperature and over current protection, sensor error detection
Incubators are constructed with a durable, powder-coated steel exterior, stainless steel interior and glass wool insulation. Units feature a 3,3 cm stainless steel ventilation cap, two doors (one solid, one glass), high temperature grade foamed silicone rubber door gaskets, over-temperature and over-current protection, and sensor error detection. A locking mode assists in preventing unintended temperature changes.
Delivery information: Supplied with two stainless steel shelves.