VWR®, Ovens with Natural Convection
Supplier: VWR
VWR®
VWRU414005-109EA 2810 SGD
VWRU414005-106 VWRU414005-109
VWR®, Ovens with Natural Convection
Ovens Standard Ovens
Ovens feature a vertical airflow pattern, easy-to-use electronic controls and port on the top for inserting monitors, they are perfect for drying samples that require a low-turbulence environment.
  • High performance three-sided heating mechanism
  • Advanced, adaptive, microprocessor control system with rotary knob resulting in superior temperature accuracy
  • Optimised inner circulation of air flow by gravity convection
  • Superior heat insulation provided by heat-resistant rubber packing and Air Flow Layer (AFL)
  • Over temperature and over current protection, sensor error detection
PT 100 temperature sensor is included for the best overall advantage in repeatability and stability over a long period. An RS232C interface for monitoring and controlling with a PC, storage function for both temperature and time, timer with alarm and error status up to 99 h 59 m with delay and continuous function, and a digital backlit LCD display are also included.
Delivery information: Supplied with two stainless steel shelves.