Would you like to log in again?
Grinding heads
Supplier: IKA
ORDER
Expand All / Collapse All
Description Type Material For Pk VWR Catalog Number Quantity
Cutting grinding head MF 10.1 Stainless steel Crushing fibrous substances e.g. paper, vegetation, plastics and material with a low specific weight. Maximum 15 mm granules 1 IKAA2870900
Supplier Article Number:: 2870900
Impact grinding head MF 10.2 Stainless steel Crushing hard brittle materials e.g. minerals or building materials up to Mohs hardness 6. Maximum 10 mm granules 1 IKAA2871000
Supplier Article Number:: 2871000
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty