Would you like to log in again?
Fluorescent viability stain, Quick-View
Supplier: VWR Chemicals
Quick-View™ fluorescent viability stain is a ready to use staining reagent for easy discrimination between live and dead mammalian cells. Supplied in an easy to use dropper bottle, the single-staining solution is a mixture of Acridine Orange for live cell identification and ethidium bromide for identification of dead cells. The Acridine Orange-stained live cells appear green and the ethidium bromide-stained dead cells appear red when visualised by fluorescence microscopy.
  • Pre-mixed, ready to use dye solution
  • Dropper bottle format minimises contact with hazardous reagents
  • Cells can be visualised immediately upon dye addition
  • Stains both suspended and adherent cells
Quick-View™ fluorescent viability stain is sensitive, fast and reliable. The cells and stain solution are simply mixed 1:1 and are immediately ready for visualisation. Washing, incubation and fixation steps are eliminated. The set-up, clean up and storage is simple and easy with minimal contact with reagents. The versatile stain can be used for both cells in suspension as well as adherent cells.
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Quick-View™ fluorescent viability stain 5 ml VWRCN600-5ML
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty