Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staining reagents

Supplier: Avantor Performance Materials



 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Aniline blue WS, wide mouth amber glass 25 g BAKRB362-03
Supplier Article Number:: B362-03
Safranine O, wide mouth amber glass 100 g BAKRU926-05
Supplier Article Number:: U926-05
Methylene blue, wide mouth amber glass, black T/E cap 25 g BAKRQ475-03
Supplier Article Number:: Q475-03
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty