Would you like to log in again?
Funnels, with short stem, DURAN®
Supplier: DWK Life Sciences
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.
  • DIN ISO 4798
ORDER
Expand All / Collapse All
Funnel O-Ø Stem O-Ø Stem length Pk VWR Catalog Number Quantity
250 mm 30 mm 175 mm 1 SCOT213516601
Supplier Article Number:: 213516601
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty