Would you like to log in again?
Laboratory bottles, narrow neck
Supplier: DWK Life Sciences
DURAN®, borosilicate glass 3.3 , amber, with or without screw cap
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Purely external application of the colour ensures unchanged DURAN® properties on the interior
  • USP Standard
Round, with graduation and DIN thread. With or without pouring ring and screw cap.
With printed retrace code (batch identification); batch certificate available at www.duran-group.com
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominal volume visible
Certifications: ISO 4796-1
ORDER
Expand All / Collapse All
With screw cap
Capacity Thread Ø ext. Height Pk VWR Catalog Number Quantity
25 ml GL 45 36 mm 70 mm 1 SCOT21806145 Unavailable
Supplier Article Number:: 21806145
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +65 6505 0760.
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty