Light meter pen, Traceable®
VWRU89094-706EA 326 SGD
VWRU89094-706
Light meter pen, Traceable®
Portable, easy to use, dual range light meter displays readings in both Lux and foot-candles. Tough ABS plastic housing with O-ring seal and membrane keypad keeps out dust and dirt, assuring long life in severe lab or harsh plant environments.
  • Reads in lux and footcandles
  • Simple operation eliminates the need to refer to instructions ; designed for intuitive, easy operation by everyone
  • High-contrast LCD digits are 1 cm (3/8") high
Traceable to NIST: to assure accuracy an individually serial-numbered Traceable® certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Delivery information: Supplied with a four AAA batteries and Traceable® Certificate.