Hotplate, Corning®
Supplier: Corning
Corning®
CORN6796-200EA 596 SGD
CORN6796-200
Hotplate, Corning®
Heaters Hotplates
Low-profile hot plates with a ceramic glass plate provide precision-controlled heating for laboratory applications. Microprocessor-controlled heating ensures consistent performance.
  • Space saving design
  • Hot top indicator light
  • Large knobs provide control for gloved fingers
Units feature large knobs and a mount for a ring stand support built into the back of the housing. A hot top indicator light blinks a warning when the top is too hot to touch, >60 °C (140 °F). All units feature a temperature range from 25 to 550 °C (77 to 1022 °F).
Certifications: UL and C-UL listed. FCC, IEC 1010, and GS certified.
Ordering information: Two year manufacturer's warranty.