Would you like to log in again?
Arsine generator systems
Supplier: Corning Life Sciences
PYREX®
This PYREX®, borosilicate glass arsine generator is designed for arsenic determination by the silver diethyldithiocarbamate photometric method. The 125 ml flask has a 24/40 inner standard taper joint and ball. Socket joint between the absorber and scrubber is a 12/2 spherical joint. Complete unit includes a clamp.
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Arsine Generator System, 125 ml 1 CORN3900
Supplier Article Number:: 3900
Component parts
Description Pk VWR Catalog Number Quantity
Absorber only 4 CORN3900-AO
Supplier Article Number:: 3900-AO
Scrubber only 4 CORN3900-SO
Supplier Article Number:: 3900-SO
Replacement polyacetyl clamp 10 CORN7721-13
Supplier Article Number:: 7721-13
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty