Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDCBP Mouse Monoclonal Antibody [clone: 2C12]

Supplier: Abnova
Type: Primary
Antigen: SDCBP
Clonality: Monoclonal
Clone: 2C12
Conjugation: Unconjugated
Host: Mouse
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µg ABNOH00006386-M01
Supplier Article Number:: H00006386-M01


Antigen symbol SDCBP
Antigen name Syndecan Binding Protein
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2C12
Host Mouse
Isotype none
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen synonyms ST1, pro-TGF-alpha cytoplasmic domain-interacting protein 18, MDA-9, Syntenin, SYCL, syndecan binding protein (syntenin), melanoma differentiation associated protein-9, SDCBP, TACIP18 , scaffold protein Pbp1 syntenin
Gene ID 6386
Cross adsorption non
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty