Laboratory hoods, Fiberglass 30
Supplier: Labconco
LBCP3030000EA 13390 SGD
LBCP3030000
Laboratory hoods, Fiberglass 30
Fume Hoods and Enclosures Chemical Fume Hoods
Compact, benchtop Fiberglass 30 laboratory hoods are designed for research, educational and clinical applications requiring small working space and efficient fume removal. The fiberglass liner is corrosion-resistant, durable, fire-resistant and light reflective. The smooth one-piece molded construction offers fewer crevices for easy cleaning.
  • Durable glacier white, dry powder epoxy-coated steel exterior with by-pass flow design
  • One-piece molded fiberglass interior and pre-set baffle with flame spread index less than 25 per ASTM E-84
  • Tempered safety glass 3/16 inch thick vertical-rising sash with epoxy-coated aluminum frame and a pivoting air foil to promote airflow
  • Rightside fixture panel is factory prepared to accept up to three service fixtures