Blood sedimentation pipette, acc. to Westergren
Supplier: Marienfeld
MARI3268062EA 75.78 SGD
MARI3268062
Blood sedimentation pipette, acc. to Westergren
In case of suspected infectious diseases the erythrocyte sedimentation rate of blood is examined by means of sedimentation pipettes according to Westergren.
  • Clear AR-Glas®
  • White graduation 0 to 200 in 1 mm
  • For micro method
Suitable for in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC.