Peqlab peqGOLD Cycle-Pure™, Agarose Gel Extraction Kit
Supplier: Avantor
VWRCK220-50RXNEA 490 SGD
VWRCK220-50RXN
Peqlab peqGOLD Cycle-Pure™, Agarose Gel Extraction Kit
Agarose gel extraction kit is a fast, spin column-based kit for the quick isolation of DNA fragments (40 bp to 100 kb) from agarose slices.
  • DNA isolation in 10 minutes
  • TAE/TBE agarose gel compatible
  • Spin column format
In just 10 minutes, ultra-pure DNA can be extracted that is ready to use in restriction enzyme digestion, labelling, ligation, transformation, in vitro transcription and sequencing protocols. All reagents are room temperature stable.
Delivery information: Kit includes Binding Buffer, 40 ml; Wash Buffer, 50 ml; spin columns, 50; and collection tubes, 50. Contains sufficient material for 50 isolations.