VWR® RNA EZ-Vision®, Loading buffer
Supplier: Avantor
VWR®
VWRCN717-2X1.5MLEA 54.7 SGD
VWRCN717-2X1.5ML
VWR® RNA EZ-Vision®, Loading buffer
Buffers Electrophoresis Buffers Loading Buffers
RNA EZ-Vision is a non mutagenic fluorescent dye for staining RNA bands resolved on denaturing agarose gels containing formaldehyde. The dye co-migrates with the RNA samples during electrophoresis and provides immediate band visualisation upon illumination with UV light. No post run staining or destaining is needed. The loading buffer includes formamide and bromophenol blue tracking dye.
  • Recommended for denaturing agarose gels with formaldehyde
  • Sensitive to 150 ng of RNA
  • Compatible with downstream applications including Northern blots
Designed for environments needing to reduce use of ethidium bromide.
Delivery information: Supplied in a 1,5X loading buffer.