These heavy-duty bags, in PE, with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic. Ideal for sterilising animal cages.
These heavy-duty bags with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic. Ideal for sterilizing animal cages.
- Clearly labeled with proper biohazard warnings
- Bright red coloured for quick identification
- Autoclavable up to 121 °C (250 °F)
- Clearly labeled with proper biohazard warnings
- Bright red coloured for quick identification
- Autoclavable up to 121°C (250°F)
Bags meet many state and local regulations.
Delivery information: Supplied with 20,3cm twist ties.
Certifications: Bags meet many state and local regulations.
Delivery information: Supplied with 20,3 cm twist ties.