Digital timer, one channel, Traceable®
VWRU46610-060EA 56 SGD
VWRU46610-060 VWRI609-0209
Digital timer, one channel, Traceable®
  • LCD display (6 digits)
  • Counts up and down for 23 hours, 59 minutes, 59 seconds
  • Memory recall automatically returns timer to last programmed setting
  • When countdown timing reaches zero, 'TIMES UP' flashes on display, alarm sounds, and timer counts up showing time elapsed since alarm
  • Clock function
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with magnet, flip-out stand, clip, battery and Traceable® certificate.