Safety wash bottles, wide neck, colour coded
VWRU16649-922EA 76.7 SGD
VWRU16649-922
Safety wash bottles, wide neck, colour coded
LDPE, with PP spray cap.
These bottles include PP closure with leakproof design for dispensing most common solvents. Easy-to-read, LDPE solvent-resistant bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place. Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.