Digital fluorescence camera, VisiCam® FL 1400
VWRI630-2668EA 6825.92 SGD
VWRI630-2668
Digital fluorescence camera, VisiCam® FL 1400
Thanks to the built-in buffer memory, it delivers a high frame rate and stable image transmission.
  • Measurement tools, time-lapse, video recording, image stitching, multi-focus with extended depth of focus
  • Fluorescence merge/split, pre-defined dye list, AOI, annotation and scale marker plus export data to Excel, CSV
  • Twain and Directshow compatible
Dynamic range and low noise for low light and fluorescence applications.
Ordering information: Includes USB cable, PC software and drivers. C-mount attachment (not included).
Delivery information: Supplied with a calibration slide and ruler.