Would you like to log in again?
Analogue hygrometer, humidity-on-a-card™, Traceable®
Supplier: VWR Collection
Traceable®
Easy-to-read cards which is ideal to use with bottles, chemicals, plastic bags, electronics, desiccators, and storage containers. Color changes from blue to lavender in increments of 10% RH.
  • Indicates both current humidity and the maximum humidity attained
  • Monitor humidity levels in shipments, optics and packaging
Certifications: Compliance with ISO/IEC 17025:2005 (1750.01); accredited by A2LA indicating traceability to standards provided by NIST.
ORDER
Expand All / Collapse All
Description Range Accuracy Resolution Pk VWR Catalog Number Quantity
Humidity on a card, 101,6×101,6 mm 0 - 100% RH ±0,5 % RH (24 °C) 10 % RH 6 VWRU15551-012
Supplier Article Number:: 15551-012
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty