VWR®, Ultra-Low Temperature Freezers, Upright Models, −86 °C
Supplier: VWR
VWR®
VWRU10160-766EA 30650 SGD
VWRU10160-732 VWRU10160-756 VWRU10160-766 VWRU10160-758 VWRU10160-768 VWRU10160-914
VWR®, Ultra-Low Temperature Freezers, Upright Models, −86 °C
Freezers Ultra Low Temperature Freezers
These freezers are designed and constructed for long-term performance and are ideal for applications where temperature control is critical. Modern design includes an easy to grasp handle for one-handed operation. Dual wheel, locking swivel castors allow mobility as well as secure positioning. Industrial-grade cabinets are constructed of high quality, heavy gauge, cold-rolled steel. The powder-coated finish is rust- and chip-resistant. Upright freezers feature a triple-point silicone door gasket, three stainless steel shelves that are fully adjustable (bottom of unit acts as fourth shelf), lockable castors, and a vacuum-relief port for easy re-entry following door closings.
  • Dual compressors
  • Microprocessor temperature control system
  • Advanced monitor/alarm system
  • Automatic voltage compensator
  • Cascade refrigeration system
  • Non CFC, non HFCF, non flammable refrigerants
  • The microprocessor temperature control system maintains constant freezer temperature, along with a sophisticated microprocessor monitor/alarm system to keep the operator informed of over-/under-temperature conditions, power failures, low battery and door ajar scenarios. LED indicates actual chamber temperature
  • Air-cooled, hermetically-sealed dual compressors work in unison and cycle on demand of the microprocessor temperature controller
  • To reduce cold air loss, units feature four inner compartment doors
  • Acoustic mufflers and sound-absorbing foam in the freezer base reduce the noise level in the lab
Temperature: −50 to −86 °C