Flow meter with regulator
VWRU55850-410EA 425 SGD
VWRU55850-410
Flow meter with regulator
This flow meter with regulator is a combination flowmeter and regulator designed to deliver an adjustable flow with accurate delivery even when upstream pressure fluctuates.
  • Captive inlet nut to protect the seat
  • Internal relief system to prevent overpressurisation
  • Colour coded pressure gauge engineered to ANSI B-40.1 specifications
Certifications: ANSI B-40.1