Would you like to log in again?
Cryoboxes, with drain slots
Supplier: VWR Collection
VWR®
Cryoboxes and dividers are constructed of durable fiberboard with a protective, moisture-repellant coating. Liquid nitrogen (LN2) freezer boxes feature slots for safe LN2 drainage and reduced LN2 consumption.
  • Lid printed with a numbered grid for superior inventory control
ORDER
Expand All / Collapse All
W×D×H ext. Compartments Pk VWR Catalog Number Quantity
132×132×51 mm 81 (9×9) 1 VWRU89214-752
Supplier Article Number:: 89214-752
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty