Power amplifiers board
VWRU470210-758EA 1290 SGD
VWRU470210-758
Power amplifiers board
The power amplifiers board is a comprehensive instructional module that plugs directly into the Electronics Universal Training System base unit designed to teach the fundamental concepts of power amplifier circuits and their related components to students who are both beginners and experts alike. The topics covered on this board include: transistor power amplifiers, class A, B, C and AB amplifiers, operational amplifier power amplifiers, and troubleshooting exercises.
A minimal amount of wiring is required to setup the board, increasing the amount of time students have for hands on learning. The fault insertion system allows you to create an error on the board for students to troubleshoot, further developing their diagnostic skills. By creating, testing, and performing experiments from the included manual, students will learn the theory, practical applications, and gain a working knowledge of the fundamental concepts of power amplifiers.