Motor and generator control board
VWRU470210-764EA 2240 SGD
VWRU470210-764
Motor and generator control board
The motor and generator control board is a comprehensive instructional module that plugs directly into the Electronics Universal Training System base unit designed to teach the fundamental concepts of stepper motors, digital to analog converters, pulse width modulation circuits, electric generators and dynamo speed controllers to students who are both beginners and experts alike. The topics covered on this board include: stepper motor control, DAC and PWM, DC motor and speed control, generators, and troubleshooting exercises. A minimal amount of wiring is required to setup the board, increasing the amount of time students have for hands on learning. The fault insertion system allows you to create an error on the board for students to troubleshoot, further developing their diagnostic skills.
  • By creating, testing, and performing experiments from the included manual, students will learn the theory, practical applications, and gain a working knowledge of the fundamental concepts of motor and generator controllers