Singapore
Order Entry
VWR®, Bottles, Wide Neck, with Screw Cap
  VWRU414004-187
undefined
VWR®, Bottles, Wide Neck, with Screw Cap
  VWRU414004-187
$36.70
  1 * 1 items
  • Thread:
    100-415
  • Capacity:
    2000 ml
  • Environmentally Preferable:
  • Pk:
    1
PP, translucent.
These bottles are specifically designed for applications that require autoclaving of large volumes. They are not leakproof and are not recommended for shipping liquids.