555, ADC and DAC circuits
VWRU470210-782EA 1320 SGD
VWRU470210-782
555, ADC and DAC circuits
The 555, ADC and DAC circuits board is a comprehensive instructional module that plugs directly into the Electronics Universal Training System base unit designed to teach the fundamental concepts of timer circuits, analog to digital conversion and digital to analog conversion circuits to students who are both beginners and experts alike. The topics covered on this board include: timing with the 555, digital to analog conversions, implementing an ADC, and troubleshooting exercises. A minimal amount of wiring is required to setup the board, increasing the amount of time students have for hands on learning. The fault insertion system allows you to create an error on the board for students to troubleshoot, further developing their diagnostic skills.
  • By creating, testing, and performing experiments from the included manual, students will learn the theory, practical applications, and gain a working knowledge of the fundamental concepts of 555, ADC and DAC circuits