This digital tachometer with unique memory allows to recall the highest, lowest and last readings, so that to check speed of centrifuges, motors and stirrers.
- Only three seconds for most readings
- Five digit LCD featuring 12,7 mm high display
- Contact readings with rotating spindle and touchless operation
Certifications: ISO/IEC 17025:2005 (1750.01) accredited by A2LA indicating traceability to NIST.
Delivery information: Supplied with batteries for one year, Traceable® certificate, plastic case, self-adhesive reflecting tabs, internal light, cone for RPM, wheel for RPM, wheel for surface speeds and carrying case.