Reagent reservoirs
VWRU80092-466EA 33.4 SGD
VWRU80092-466
Reagent reservoirs
PP, translucent. These chemical resistant reservoirs are compatible with single, 8-, or 12-channel pipettors.
  • Minimising waste, V-shaped wells allow complete sample withdrawal
  • Each channel holds 5 ml, with numbers to facilitate serial dilution
  • Wide profile provides added stability
  • Individually sealed and sterilised; autoclavable at 121 °C (250 °F)
  • Fitted lid included