Liebig condenser with standard ground joints, KIMBLE®
Supplier: DWK Life Sciences
KIMAX®
KIML447000-2425EA 391 SGD
KIML447000-2425
Liebig condenser with standard ground joints, KIMBLE®
Condensers
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Liebig condensers with hose tubing connectors (accept 9,5 mm int. Ø flexible tubing), standard taper outer joint at the top and standard taper inner drip joint at the bottom.
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.