Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulphurous acid (≥ 6,0% SO₂) in aqueous solution , AR® ACS

Supplier: Avantor Performance Materials
Danger
Synonyms: Sulphur dioxide solution



Formula: H₂O₃S
MW: 82,08 g/mol
Density: 1,03 g/cm³ (20 °C)
MDL Number: MFCD00011335
CAS Number: 7782-99-2
EINECS: 231-973-1
UN: 1833
ADR: 8,II
Specification Test Results
ACS - Assay (as SO₂) >= 6.0 %
ACS - Chloride (Cl) <= 5 ppm
ACS - Heavy Metals (as Pb) <= 2 ppm
ACS - Iron (Fe) <= 5 ppm
ACS - Residue after Ignition <= 0.005 %

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
clear glass bottle 2,5 l BAKR2920-46
Supplier Article Number:: 2920-46
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty