Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl acetate ≥98%

Supplier: Avantor Performance Materials
Danger



Formula: C₃H₆O₂
MW: 74,08 g/mol
Boiling Pt: 56,9 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0,938 g/cm³ (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3,II
Merck Index: 12,06089
Specification Test Results
Assay (CH₃COOCH₃) (by GC) >= 98 %

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
NMA glass btl/single shipper 500 ml BAKRQ007-07
Supplier Article Number:: Q007-07
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty