Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead (II) acetate trihydrate 99.0-103.0% BAKER ANALYZED® ACS

Supplier: Avantor Performance Materials
Danger

Granular. Lot analysis on label.


Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379,34 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C
Density: 2,55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 13,05415
https://sg.vwr.com/stibo/web/std.lang.all/33/36/713336.jpg
Specification Test Results
Meets ACS Reagent Chemical Requirements
Assay ((CH₃COO)₂Pb3H₂O) (by EDTA titrn) 99.0 - 103.0 %
Insoluble Matter <= 0.01 %
pH of 5% Solution at 25°C 5.5 - 6.5
Nitrate and Nitrite (as NO₃) <= 0.005 %
Calcium (Ca) <= 0.005 %
Copper (Cu) <= 0.002 %
Trace Impurities - Iron (Fe) <= 0.001 %
Potassium (K) <= 0.005 %
Sodium (Na) <= 0.010 %
Chloride (Cl) <= 5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
- 110 lb BAKR2271-08
Supplier Article Number:: 2271-08
Poly Bottle 500 g BAKR2271-01
Supplier Article Number:: 2271-01
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty