Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formamide

Supplier: Avantor Performance Materials
Danger
Synonyms: Carbamaldehyde



Formula: HCONH₂
MW: 45,04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1,1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
UN: 0000
Merck Index: 13,04261
https://sg.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg
Specification Test Results
Freezing Point <= 3.5 °C

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
NMA glass btl/single shipper 500 ml BAKRM520-07
Supplier Article Number:: M520-07
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty