Mercury(II) acetate ≥98.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Acetic acid mercury (II) saltMercury diacetateMercuric acetate

BAKR2584-04EA 132 SGD
BAKR2584-04
Mercury(II) acetate ≥98.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
MW: 318,68 g/mol
Melting Pt: 178…180 °C
Density: 3,27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃COO)₂Hg) ≥98.0 %
Insoluble Matter ≤0.01 %
Residue after Reduction ≤0.020 %
Chloride (Cl) ≤0.005 %
Nitrate (NO₃) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Other Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Mercurous Mercury (as Hg) ≤0.4 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR