Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycolic acid ≥98%

Supplier: Avantor Performance Materials
Danger
Synonyms: 2-Hydroxy acid

Crystals.


Formula: C₂H₄O₃
MW: 76,05 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1,49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511
Specification Test Results
Assay (C₂H₄O₃) (by acid-base titration) >= 98 %
Appearance Passes Test

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
wide mouth amber glass 100 g BAKRM821-05
Supplier Article Number:: M821-05
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty