Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetanilide BAKER ANALYZED®

Supplier: Avantor Performance Materials
Warning

Lot analysis on label. Reference standard for determination of carbon, hydrogen, nitrogen, and oxygen.


Formula: C₈H₉NO
MW: 135,17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1,22 g/cm³ (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
UN: 0000
Merck Index: 13,00051
Specification Test Results
Carbon (C) For Information Only
Hydrogen (H) For Information Only
Nitrogen (N) For Information Only
Oxygen (O) For Information Only
Appearance (white to off-white, grayish or brownish crystalline scales or crystalline powder)

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
wide mouth amber glass 25 g BAKRA068-03
Supplier Article Number:: A068-03
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping